6-Methoxy-2-naphthoic acid 97%

Supplier: Ambeed
Warning

A146840-100G A146840-10G A146840-1G A146840-25G A146840-5G
103803-704EA 12 USD
103803-704 103841-798 76997-106 76997-108 77317-136 77317-138 77317-140
6-Methoxy-2-naphthoic acid 97%
6-Methoxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Boiling Pt: 371.1 °C (760 mm Hg)
Melting Pt: 192…194 °C
Flash Pt: 147.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00092750
CAS Number: 2471-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR