6-Methoxy-2-methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-methylpyridin-3-amine, 3-Amino-6-methoxy-2-methylpyridine, 3-Amino-6-methoxy-2-picoline

A135419-100G A135419-10G A135419-250MG A135419-5G A135419-25G
103802-150EA 17 USD
103802-150 77095-184 77095-186 77095-188 77298-778 77314-966
6-Methoxy-2-methylpyridin-3-amine 98%
6-Methoxy-2-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1671 g/mol
Boiling Pt: 260 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03095090
CAS Number: 52090-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR