6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-(4-methoxyphenyl)-1-benzothiophene

A162077-100G A162077-10G A162077-5G A162077-1G A162077-25G
103805-570EA 6 USD
103805-570 103843-818 76812-808 76812-810 77300-572 77319-848 77319-850
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene 95%
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂S
MW: 270.35 g/mol
Boiling Pt: 435.7 °C (760 mm Hg)
Melting Pt: 191…197 °C
Storage Temperature: Ambient
MDL Number: MFCD02083138
CAS Number: 63675-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR