6-Methoxy-1-naphthoic acid 95%

Supplier: Ambeed
Warning

A146852-100MG A146852-250MG A146852-1G A146852-5G
103803-712EA 29 USD
103803-712 103841-806 76768-014 76768-016 77317-154 77317-156
6-Methoxy-1-naphthoic acid 95%
6-Methoxy-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.2 g/mol
Boiling Pt: 403.3 °C (760 mm Hg)
Flash Pt: 162.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00090331
CAS Number: 36112-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR