6-Methoxy-1H-indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A126326-100MG A126326-10G A126326-250MG A126326-1G A126326-25G A126326-5G
103800-794EA 12 USD
103800-794 77026-826 77026-828 77026-830 77313-002 77313-004 77313-006
6-Methoxy-1H-indole-3-carbaldehyde 98%
6-Methoxy-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 375.2 °C (760 mm Hg)
Flash Pt: 180.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03848058
CAS Number: 70555-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR