6-Methoxy-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyindole-2-carboxylic acid

A147793-250MG A147793-5G
103803-834EA 30 USD
103803-834 103841-942 77272-850 77317-346
6-Methoxy-1H-indole-2-carboxylic acid 98%
6-Methoxy-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Boiling Pt: 447.6 °C (760 mm Hg)
Melting Pt: 198…203 °C
Flash Pt: 224.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01548823
CAS Number: 16732-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR