5-Methoxyisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione, 6-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione

A103324-250MG A103324-25G A103324-1G A103324-5G
103796-530EA 34 USD
103796-530 103822-234 77257-350 77257-352 77283-076 77309-106
5-Methoxyisatoic anhydride 97%
5-Methoxyisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179411
CAS Number: 37795-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR