6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Supplier: Ambeed
Warning

A128177-100G A128177-10G A128177-250MG A128177-1G A128177-25G A128177-5G
103801-078EA 15 USD
103801-078 77161-724 77161-726 77161-728 77313-364 77313-366 77313-368
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
Melting Pt: 230…232.5 °C
Flash Pt: 109.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01717328
CAS Number: 57196-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR