6-Methoxy-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

A313694-100MG A313694-250MG A313694-1G A313694-5G
103825-018EA 39 USD
103825-018 103835-930 77167-168 77167-170 77306-098 77306-100
6-Methoxy-1,2,3,4-tetrahydroisoquinoline 97%
6-Methoxy-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.2163 g/mol
Boiling Pt: 283.615 °C (760 mm Hg)
Flash Pt: 109.208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01717129
CAS Number: 42923-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR