EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester) 98%
Supplier: Ambeed
|
Warning
|
A167523-100MG
A167523-10G
A167523-1MG
A167523-50MG
A167523-25G
A167523-5G
A167523-100G
A167523-250MG
103806-196EA
12
USD
103806-196
103844-456
76799-264
76799-266
76799-268
76799-270
77081-472
77081-474
77600-348
77879-122
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester) 98%
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester)
Precaución: for Research Use Only
Formula:
C₁₄H₁₆N₂O₆ Boiling Pt: 480.6 °C (760 mm Hg) Melting Pt: 70…73 °C Flash Pt: 244.5 °C Storage Temperature: Freezer |
MDL Number:
MFCD00043043 CAS Number: 55750-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...