6-Iodoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Iodo-1-azanaphthalene

A355790-1G A355790-25G A355790-5G A355790-10G A355790-250MG
103826-150EA 12 USD
103826-150 103837-158 76840-284 76840-286 76840-288 77115-192 77115-194
6-Iodoquinoline 98%
6-Iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₆IN
MW: 255.0600 g/mol
Boiling Pt: 320.7 °C (760 mm Hg)
Melting Pt: 86…90 °C
Flash Pt: 147.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08457906
CAS Number: 13327-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR