6-Iodoquinazolin-4-one 97%

Supplier: Ambeed
Warning

A149373-100G A149373-1G A149373-25G A149373-5G
103804-034EA 6 USD
103804-034 103842-152 76768-092 77317-640 77317-642 77317-644
6-Iodoquinazolin-4-one 97%
6-Iodoquinazolin-4-one

Precaución: for Research Use Only

Formula: C₈H₅IN₂O
MW: 272.04 g/mol
Boiling Pt: 401 °C (760 mm Hg)
Melting Pt: 272…274 °C
Flash Pt: 196.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05662719
CAS Number: 16064-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR