6-Iodopyridazin-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-6-iodopyridazine

A445210-100MG A445210-10G A445210-250MG A445210-1G A445210-25G A445210-5G
103827-728EA 17 USD
103827-728 103838-792 76818-264 76818-266 76818-268 76842-578 76842-580 76842-582
6-Iodopyridazin-3-amine 95%
6-Iodopyridazin-3-amine

Precaución: for Research Use Only

Formula: C₄H₄IN₃
MW: 221 g/mol
Boiling Pt: 399.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09991754
CAS Number: 187973-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR