6-Iodo-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Iodoindazole

A173590-1G A173590-5G A173590-100G A173590-10G A173590-250MG
103806-834EA 9 USD
103806-834 103845-134 77082-382 77082-384 77163-648 77163-650 77163-652
6-Iodo-1H-indazole 98%
6-Iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.0300 g/mol
Boiling Pt: 358.2 °C (760 mm Hg)
Flash Pt: 170.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114695
CAS Number: 261953-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR