6-Quinolinol 97%

Supplier: Ambeed
Warning

A153387-100G A153387-10G A153387-1G A153387-25G A153387-5G
103804-504EA 5 USD
103804-504 103842-676 76689-450 76689-452 77300-006 77318-330 77318-332
6-Quinolinol 97%
6-Quinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00047611
CAS Number: 580-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR