6-Hydroxypyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A311789-1G A311789-25G A311789-5G A311789-100G A311789-100MG A311789-10G A311789-250MG
103814-680EA 8 USD
103814-680 103835-870 76838-674 76838-676 76838-678 77015-156 77015-158 77015-160 77015-162
6-Hydroxypyrimidine-4-carboxylic acid 97%
6-Hydroxypyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
Boiling Pt: 250.8 °C (760 mm Hg)
Melting Pt: 159 °C
Flash Pt: 138.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023267
CAS Number: 6299-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR