6-Hydroxypyridine-2-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxypicolinamide

A283039-1G A283039-100MG A283039-250MG A283039-5G
103813-600EA 100 USD
103813-600 76837-254 77194-194 77194-196 77194-198
6-Hydroxypyridine-2-carboxamide 97%
6-Hydroxypyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506698
CAS Number: 89640-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR