6-Hydroxyindole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A127981-250MG A127981-1G A127981-5G
103801-048EA 76 USD
103801-048 77094-854 77313-332 77313-334
6-Hydroxyindole-2-carboxylic acid 95%
6-Hydroxyindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01549019
CAS Number: 40047-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR