6-Hydroxyhexanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxycaproic acid

A247480-100MG A247480-10G A247480-250MG A247480-1G A247480-25G A247480-5G A247480-100G
103811-770EA 20 USD
103811-770 103851-606 76647-734 76647-736 76647-738 77304-408 77304-410 77304-412 77616-028
6-Hydroxyhexanoic acid 95%
6-Hydroxyhexanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Boiling Pt: 272.6 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 132.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00046560
CAS Number: 1191-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR