6-Hydroxybenzothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 6-Benzothiazolol

A116184-10G A116184-1G A116184-5G A116184-250MG A116184-25G
103798-688EA 36 USD
103798-688 103824-552 76766-206 77297-434 77297-436 77570-992 77599-678
6-Hydroxybenzothiazole 95%
6-Hydroxybenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅NOS
MW: 151.1857 g/mol
Boiling Pt: 311.6 °C (760 mm Hg)
Flash Pt: 142.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00057883
CAS Number: 13599-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR