6-Hydroxy-3-coumaranone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one

A163425-100G A163425-10G A163425-1G A163425-5G
103805-724EA 11 USD
103805-724 103843-984 77235-952 77235-954 77320-080 77320-082
6-Hydroxy-3-coumaranone 98%
6-Hydroxy-3-coumaranone

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.14 g/mol
Boiling Pt: 369.3 °C (760 mm Hg)
Melting Pt: 242…246 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00068174
CAS Number: 6272-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR