6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-hydroxybenzo[d]thiazole-2-carbonitrile, 2-Cyano-6-hydroxybenzothiazole, 6-hydroxy-1,3-benzothiazole-2-carbonitrile

A188291-1G A188291-25G A188291-5G A188291-10G A188291-250MG A188291-100MG
103816-086EA 15 USD
103816-086 103846-678 77084-402 77084-404 77084-406 77260-450 77260-452 77748-212
6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
6-Hydroxybenzo[d]thiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄N₂OS
Boiling Pt: 374.7±34.0 °C (760 mm Hg)
Melting Pt: 212…215 °C
Flash Pt: 180.387 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00296905
CAS Number: 939-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR