6-Cyano-2-naphthol 97%

Supplier: Ambeed
Warning

A163818-100G A163818-10G A163818-1G A163818-25G A163818-5G
103805-772EA 9 USD
103805-772 103844-036 76799-110 76799-112 77300-678 77320-152 77320-154
6-Cyano-2-naphthol 97%
6-Cyano-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₇NO
Melting Pt: 165.5…170.5 °C
Flash Pt: 185.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01863480
CAS Number: 52927-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR