6-Hydroxy-2-naphthoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid

A127397-1000G A127397-100G A127397-10G A127397-500G A127397-5G A127397-25G
76797-728EA 84.88 USD
76797-728 76797-730 76797-732 76797-734 77298-206 77313-204
6-Hydroxy-2-naphthoic acid 98%
6-Hydroxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Boiling Pt: 407 °C (760 mm Hg)
Melting Pt: 240…250 °C
Flash Pt: 214.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00060070
CAS Number: 16712-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR