6-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A820369-250MG A820369-5G
103841-128EA 1127 USD
103841-128 77019-520 77308-386
6-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 97%
6-Hydroxy-2-methyl-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13179449
CAS Number: 308110-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR