6-Hydroxy-1-tetralone 97%

Supplier: Ambeed
Warning

A243720-100G A243720-10G A243720-25G A243720-5G A243720-1G A243720-500G
103811-538EA 5 USD
103811-538 103851-364 76976-530 76976-532 76976-534 77090-572 77304-276 77600-962
6-Hydroxy-1-tetralone 97%
6-Hydroxy-1-tetralone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Melting Pt: 154…157 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00156670
CAS Number: 3470-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR