6-Hydroxy-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydroquinolin-6-ol

A126953-10G A126953-250MG A126953-1G A126953-25G A126953-5G
103800-882EA 12 USD
103800-882 77094-814 77094-816 77313-122 77313-124 77313-126
6-Hydroxy-1,2,3,4-tetrahydroquinoline 95%
6-Hydroxy-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.1897 g/mol
Boiling Pt: 337.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00800243
CAS Number: 3373-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR