6H-Benzo[c]chromen-6-one 96%

Supplier: Ambeed
Warning

A214001-1G A214001-5G
103818-356EA 99 USD
103818-356 103849-066 77303-316 77303-318
6H-Benzo[c]chromen-6-one 96%
6H-Benzo[c]chromen-6-one

Precaución: for Research Use Only

Formula: C₁₃H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00027376
CAS Number: 2005-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR