6-Formylpicolinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-Formylpyridine-2-carbonitrile

A100615-100MG A100615-250MG A100615-1G A100615-5G
103795-538EA 81 USD
103795-538 103821-228 76869-104 76869-106 77282-520 77282-522
6-Formylpicolinonitrile 97%
6-Formylpicolinonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O
MW: 132.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01108768
CAS Number: 85148-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR