6-Fluoroquinoline 97%
Supplier: Ambeed
|
Warning
|
A131101-10G
A131101-1G
A131101-5G
103801-524EA
10
USD
103801-524
76782-350
77314-002
77314-004
6-Fluoroquinoline 97%
6-Fluoroquinoline
Precaución: for Research Use Only
Formula:
C₉H₆FN MW: 147.15 g/mol Boiling Pt: 238.4 °C (760 mm Hg) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD01685512 CAS Number: 396-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...