6-Fluoroquinazolin-4(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoroquinazolin-4-one

A455984-10G A455984-250MG A455984-1G A455984-25G A455984-5G
103827-852EA 21 USD
103827-852 103838-926 76774-160 76774-162 76842-770 76842-772 76842-774
6-Fluoroquinazolin-4(3H)-one 98%
6-Fluoroquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MW: 164.14 g/mol
Boiling Pt: 318.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09954859
CAS Number: 16499-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR