6-Fluoropyrimidine-2,4-diamine 95%

Supplier: Ambeed
Warning

A448235-1G A448235-100MG
103827-774EA 79 USD
103827-774 103838-840 76842-652 77291-662
6-Fluoropyrimidine-2,4-diamine 95%
6-Fluoropyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₄H₅FN₄
Boiling Pt: 408.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00219492
CAS Number: 696-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR