6-Fluoropicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid

A143604-100G A143604-10G A143604-1G A143604-25G A143604-5G
103803-304EA 12 USD
103803-304 76972-712 76972-714 77316-590 77316-592 77316-594
6-Fluoropicolinic acid 98%
6-Fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 306.3 °C (760 mm Hg)
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD02181193
CAS Number: 402-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR