2-Fluoro-5-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-pyridinol, 6-Fluoropyridin-3-ol

A219897-100G A219897-100MG A219897-10G A219897-250MG A219897-1G A219897-25G A219897-5G
103818-826EA 11 USD
103818-826 103849-708 76975-798 76975-800 76975-802 76975-804 77088-286 77088-288 77088-290
2-Fluoro-5-hydroxypyridine 98%
2-Fluoro-5-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
Boiling Pt: 319.8 °C (760 mm Hg)
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD01075040
CAS Number: 55758-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR