2-Fluoro-6-formylpyridine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoropicolinaldehyde, 6-Fluoropyridine-2-carboxaldehyde
A157871-100MG
A157871-250MG
A157871-1G
A157871-5G
A157871-10G
103805-056EA
25
USD
103805-056
103843-262
76997-650
76997-652
77319-098
77319-100
77567-294
2-Fluoro-6-formylpyridine 95%
2-Fluoro-6-formylpyridine
Precaución: For research use only.
Formula:
C₆H₄FNO MW: 125.1 g/mol Boiling Pt: 192.4 °C (760 mm Hg) Flash Pt: 126.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095149 CAS Number: 208110-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...