6-fluoronicotinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-2-fluoropyridine, 6-Fluoronicotinonitrile

A251861-1G A251861-5G A251861-10G A251861-250MG A251861-25G
103812-012EA 6 USD
103812-012 103851-852 77091-248 77091-250 77113-468 77113-470 77304-566
6-fluoronicotinonitrile 97%
6-fluoronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.102 g/mol
Boiling Pt: 223.7 °C (760 mm Hg)
Flash Pt: 89.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09261087
CAS Number: 3939-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR