6-Fluoronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-pyridinecarboxylic acid

A151605-10G A151605-25G A151605-5G A151605-1G A151605-100G
103804-280EA 5 USD
103804-280 103842-432 77095-826 77299-886 77299-888 77318-010 77685-260
6-Fluoronicotinic acid 98%
6-Fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 309.4 °C (760 mm Hg)
Melting Pt: 144…148 °C
Flash Pt: 140.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01859863
CAS Number: 403-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR