6-Fluoro-2,3-dihydro-1H-isoindol-1-one 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoro-1-isoindolinone, 6-Fluoroisoindolin-1-one
A130689-100MG
A130689-250MG
A130689-1G
A130689-5G
103801-460EA
107
USD
103801-460
76811-152
76811-154
77313-914
77313-916
6-Fluoro-2,3-dihydro-1H-isoindol-1-one 95%
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Precaución: for Research Use Only
Formula:
C₈H₆FNO MW: 151.138 g/mol Boiling Pt: 380.4±42.0 °C (760 mm Hg) Flash Pt: 183.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09701283 CAS Number: 340702-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...