6-Fluorooxindole 98%

Supplier: Ambeed
Warning

A235178-1G A235178-25G A235178-5G A235178-10G A235178-250MG
103810-992EA 9 USD
103810-992 103850-756 77089-756 77089-758 77089-760 77275-836 77275-838
6-Fluorooxindole 98%
6-Fluorooxindole

Precaución: for Research Use Only

Formula: C₈H₆FNO
Boiling Pt: 287.5 °C (760 mm Hg)
Melting Pt: 132…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047212
CAS Number: 56341-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR