6-Fluoro-2(3H)-benzothiazolone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluorobenzo[d]thiazol-2(3H)-one

A274030-100MG A274030-250MG A274030-1G A274030-5G
103813-174EA 26 USD
103813-174 103853-098 76816-268 76816-270 77092-912 77092-914
6-Fluoro-2(3H)-benzothiazolone 98%
6-Fluoro-2(3H)-benzothiazolone

Precaución: for Research Use Only

Formula: C₇H₄FNOS
Melting Pt: 189…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01663356
CAS Number: 63754-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR