6-Fluorobenzo[d]isoxazol-3-ylamine 95%

Supplier: Ambeed
Warning

A107113-250MG A107113-1G A107113-100MG
103797-128EA 306 USD
103797-128 103822-886 76780-886 77283-540 77599-552
6-Fluorobenzo[d]isoxazol-3-ylamine 95%
6-Fluorobenzo[d]isoxazol-3-ylamine

Precaución: for Research Use Only

Formula: C₇H₅FN₂O
MW: 152.1300 g/mol
Boiling Pt: 315 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09999138
CAS Number: 177995-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR