6-Fluorobenzo[d]isoxazol-3(2H)-one 95%

Supplier: Ambeed
Warning

A133925-100MG A133925-250MG
103801-932EA 133 USD
103801-932 76823-152 76823-154
6-Fluorobenzo[d]isoxazol-3(2H)-one 95%
6-Fluorobenzo[d]isoxazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₄FNO₂
MW: 153.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13180537
CAS Number: 99822-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR