5-Amino-2-fluoro-3-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-fluoro-3-picoline, 6-Fluoro-5-methylpyridin-3-amine
A441516-1G
A441516-250MG
103827-674EA
39
USD
103827-674
103838-744
76842-508
77004-268
5-Amino-2-fluoro-3-methylpyridine 97%
5-Amino-2-fluoro-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Boiling Pt: 275.5 °C (760 mm Hg) Melting Pt: 117…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03095088 CAS Number: 186593-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...