6-Fluoro-4-methylnicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Carboxy-2-fluoro-4-methylpyridine, 6-Fluoro-4-methylpyridine-3-carboxylic acid, 2-Fluoro-4-methylpyridine-5-carboxylic acid

A122663-250MG A122663-1G
103800-250EA 18 USD
103800-250 76983-328 77312-380
6-Fluoro-4-methylnicotinic acid 97%
6-Fluoro-4-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09260452
CAS Number: 944582-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR