6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Paliperidone Related Compound B

A153776-100G A153776-5G A153776-500G
103842-742EA 17 USD
103842-742 76719-824 77318-424 77667-170
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride 98%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClFN₂O
MW: 256.7000 g/mol
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD06658529
CAS Number: 84163-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR