6-Fluoroquinaldine 98%

Supplier: Ambeed
Warning

A107285-100G A107285-10G A107285-250MG A107285-25G A107285-1G A107285-5G
103797-160EA 9 USD
103797-160 103822-926 76765-554 76765-556 76765-558 76765-560 77309-774 77309-776
6-Fluoroquinaldine 98%
6-Fluoroquinaldine

Precaución: for Research Use Only

Formula: C₁₀H₈FN
MW: 161.18 g/mol
Melting Pt: 49…53 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041233
CAS Number: 1128-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR