6-Fluoro-2-methylnicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-methyl-3-pyridinecarboxylic acid

A192524-1G A192524-25G A192524-5G A192524-250MG
103816-450EA 31 USD
103816-450 103847-058 77084-930 77084-932 77084-934 77111-836
6-Fluoro-2-methylnicotinic acid 97%
6-Fluoro-2-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.1300 g/mol
Flash Pt: 133.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05662400
CAS Number: 884494-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR