6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline 98%
Supplier: Ambeed
|
Warning
|
A230495-100MG
A230495-250MG
A230495-1G
103819-550EA
36
USD
103819-550
103850-450
76976-128
76976-130
77089-312
6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline 98%
6-Fluoro-2-methyl-1,2,3,4-tetrahydroquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₁₂FN MW: 165.2100 g/mol Boiling Pt: 254.6 °C (760 mm Hg) Melting Pt: 31…33 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040976 CAS Number: 42835-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...