Brequinar 96%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-(2'-fluoro-[1,1'-biphenyl]-4-yl)-3-methylquinoline-4-carboxylic acid

A155500-100MG A155500-250MG
103791-498EA 45 USD
103791-498 103792-218 103804-770 76689-538 76689-540
Brequinar 96%
Brequinar

Precaución: for Research Use Only

Formula: C₂₃H₁₅F₂NO₂
MW: 375.37 g/mol
Boiling Pt: 550.9 °C (760 mm Hg)
Flash Pt: 287 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866437
CAS Number: 96187-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR