6-Fluoro-7-azaindole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1H-pyrrolo[2,3-b]pyridine

A119040-250MG A119040-100MG
103799-154EA 163 USD
103799-154 77571-126 77610-992
6-Fluoro-7-azaindole 97%
6-Fluoro-7-azaindole

Precaución: Sealed in dry, at 2…8°C

Formula: C₇H₅FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD06659668
CAS Number: 898746-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR